Learn More
Heidolph™ Multi-channel Pump Head
Increase your throughput by running more cassettes on one single pump drive and pump head system
Brand:  Heidolph™ 524-40810-00
 View more versions of this product
Product Code. 10504742
  • 1585.00€ / Each
Estimated Shipment: 31-01-2025
to see stock.
Explore more special offers
Add to basket
Récupel-bijdrage uitgesloten
Description
Description
  • Separate metering into multiple vessels with different feed rates in a single operation by using different tubing dimensions to adjust flow rate
  • Save time - tubing change is done in just seconds
  • Change your cassettes easily, even during operation – there are no restrictions
  • Cassette adjustments and changes are simple, just click in place
  • No additional purchase of a pump drive needed: upgrade your single-channel pump drive with just one adaptor for multi-channel use and pick the head/cassette configuration that matches your application needs
Specifications
Specifications
8 medium cassettes
4 rollers
C 8
Pump Head