Learn More
Nabertherm™ Muffle Furnaces with Brick Insulation and Lift Door
For research and laboratory applications.
Brand:  Nabertherm™ L-155H3ANO
Product Code. 17337899
  • 7115.00€ / Each
Estimated Shipment: 20-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Heating from two sides
  • Heating elements on support tubes ensure free heat radiation and a long service life
  • Multi-layer insulation with robust lightweight refractory bricks in the furnace chamber
  • Housing made of sheets of textured stainless steel
  • Dual shell housing for low external temperatures and stability
  • Lift door (LT) with hot surface facing away from operator
  • Adjustable air inlet in the furnace door
  • Exhaust air outlet in rear wall of furnace
  • Solid state relays provide for low-noise operation
  • NTLog Basic for Nabertherm controller: recording of process data with USB-flash drive
Additional equipment
  • Chimney, chimney with fan or catalytic converter
  • Over-temperature limiter with adjustable cutout temperature for thermal protection class 2 in accordance with EN 60519-2 as temperature limiter to protect the furnace and load
  • Protective gas connection to purge with non-flammable protective or reaction gases
  • Manual or automatic gas supply system
Specifications
Specifications
C550
3.5 kw
530 mm
625 mm
15 L
340 x 260 x 170 mm
630 mm