Learn More
Tocris Bioscience™ MRS 2578
Selective P2Y6 antagonist
Brand:  Tocris Bioscience™ 2146/50
534.74 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18004492
  • 673.00€ / 50mg
Explore more special offers
Description
Description
MRS 2578 is a selective antagonist of P2Y6 nucleotide receptors; IC50 values are 37 and 98 nM at human and rat P2Y6 receptors respectively. Displays no activity at P2Y1, P2Y2, P2Y4 and P2Y11 receptors (IC50 > 10 μM). Inhibits agonist-induced cardiomyocyte contraction and UDP-induced phagocytosis.
Specifications
Specifications
50 mg
MRS 2578
472.66
MRS 2578