missing translation for 'onlineSavingsMsg'
Learn More

Heidolph™ MR Hei-Tec Magnetic Stirrer with Heating

Product Code. 10690555 Shop All Heidolph Instruments Products
Click to view available options
Temperature (Metric):
300°C (Max.)
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10690555

Brand: Heidolph™ 5053000000

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Includes leading safety standards and features for superior ease of use and reduced cost of ownership. Heidolph™ MR Hei-Tec Magnetic Stirrer with Heating includes a variety of features for easy monitoring and control.

  • Digital display for easy setting and monitoring.
  • Independent safety circuit that switches off heating if hotplate temperature exceeds 25°C over set temperature.
  • Digital speed and temperature setting.
  • Illuminated stirring mode button.
  • Illuminating on/off switch for heating to prevent unintentional heat-up.
  • Residual heat indicator to prevent burning hazards when heat is switched off.
  • Upgradable with optional temperature sensor for precise temperature control, overshoot protection and reproducible results.
  • Safety circuit that switches off heating if the optional temperature sensor is not immersed in your media vessel.
TRUSTED_SUSTAINABILITY

Specifications

Temperature (Metric) 300°C (Max.)
Type Magnetic Stirring Hotplate
Stirring Range 100 to 1400 rpm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.