missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ MPure™ Cultured Cell DNA Extraction Kit

For extraction of genomic DNA from cultured cells in monolayer or suspension and from buffy coat

Brand:  MP Biomedicals™ 117022500


Product Code. 15759210

  • 456.00€ / 1 set

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Completely automated purification protocol
  • Run times as little as 40 minutes
  • DNA can be used immediately in downstream applications such as PCR, qPCR, Sequencing (NGS), Microarray, RFLP and Southern Blot Analysis

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store at room temperature
Magnetic Bead
MPure-12 Automated Nucleic Acid Purification System
Proteinase K solution, 40μL; Lysis Buffer 2, 720μL; Binding Buffer 1, 720μL; Magnetic Bead Solution, 800μL; Washing Buffer 1, 1000μL; Washing Buffer 2, 1000μL; Washing Buffer 3, 1000μL; Elution Buffer 1, 1000μL; Elution Buffer 2, 1000μL
Do not use more than 5 x 106 cells with a normal set of chromosomes
Cultured Cell DNA Extraction Kit
Cell Cultures, Buffy Coat
DNA Purification
48 Preps
As little as 40 minutes
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • MPure Cultured Cell DNA Extraction Kit

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : Proteinase K

SDS
Documents

Documents

Product Certifications
Promotions

Promotions