missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Whole Serum, Mouse, Purified, 2mL

Whole Mouse Serum with buffer salts

Brand:  MP Biomedicals™ 0855989

Product Code. 17377459

  • 76.00€ / 2mL
Estimated Shipment: 20-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

TRUSTED_SUSTAINABILITY
Specifications

Specifications

2° to 8°C
Lyophilized
2 mL
For use as a Serum Protein control in immunoelectrophoresis, Ouchterlony immunodiffusion, and other immunoprecipitation techniques
57.3 mg/mL
Mouse
Mouse Serum
Dialyzed into Sodium Phosphate 0.02M, Sodium Chloride 0.14M, pH 7.3, Sodium Azide 0.05%
Yes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions