missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Whole Serum, Goat, Purified, 2mL
Whole Goat Serum with buffer salts
Brand: MP Biomedicals™ 0855984
121.33 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
2° to 8°C | |
Goat Serum | |
0.2 μm filtration | |
Blocking assays, immunohistochemical, immunocytochemical, immunofluorescent applications | |
63.3 mg/mL |
Goat | |
2 mL | |
Delipidated, lyophilized from PBS with Azide | |
Yes |