missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Sheep Serum

Sterile, filtered, Lyophilized Serum

Brand:  MP Biomedicals™ 08642951

Product Code. 11443976

  • 103.77€ / 50mL
Estimated Shipment: 10-09-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

TRUSTED_SUSTAINABILITY
Specifications

Specifications

-20°C
Lyophilized
500 mL
Propagation of long-term cultures of erythrocytic stages of the Theileria uilenbergi intracellular protozoan parasite
Sheep
Sheep Serum
7.2g/dL
Yes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions