missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Fibronectin from Bovine Plasma
Involved in cell adhesion to extracellular matrices, blood coagulation and wound healing
Brand: MP Biomedicals™ 0215002505
776.10 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Format: White fiberous material
- Purity: ≥90%
- Molecular weight: 220,000d
- Sterile filtered
Dentistry; Cell Biology and Analysis
Specifications
0.5 to 50μg/mL | |
220000 | |
5 mg | |
4°C |
Cell Biology and Analysis | |
Fibronectin from Bovine Plasma | |
Bovine Plasma | |
Lyophilized |