missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Deoxyribonuclease I from Bovine Pancreas
Shop All MP Biomedicals Products
Click to view available options
Quantity:
5 mg
20 mg
Unit Size:
20mg
5mg
Description
An endonuclease that splits phosphodiester linkages, preferentially adjacent to a pyrimidine nucleotide. The average chain of limit digest is a tetranucleotide. DNase I acts upon single chain DNA, and upon double-stranded DNA and chromatin.
- CAS Number: 9003-98-9
- EC Number: 232-667-0
- MDL Number: MFCD00130918
- Format: White powder
Specifications
Specifications
| Concentration | ≥2000 kU/mg |
| Color | White |
| pH | 7.8 |
| For Use With (Application) | Used as a genetic marker for forensic identification |
| Content And Storage | 4°C |
| Source | Bovine pancreas |
| Quantity | 20 mg |
| Product Type | Deoxyribonuclease I |
| Molecular Weight (g/mol) | 31000 |
| Inhibitors | EDTA 7,8 and Sodium Dodecyl Sulfate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction