missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ D-(+)- RAFFINOSE PENTAHYDRATE (POWDER)

Crystalline white, pure (≥98%) raffinose pentahydrate powder with applications in the areas of yeast cell culture and gene expression

Brand:  MP Biomedicals™ 114061522

Product Code. 11387839

  • 291.00€ / 100g

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Purity:≥98%
  • Physical Description: White Crystalline Powder
  • Formula: C18H32O165H2O
  • Formula Weight: 594.5
  • CAS #:17629-30-0
  • FTIR: Conforms to Standard
  • Melting Point: 76- 86°C
  • Optical Rotation (c=4, water): 101-107°
  • Loss on Drying: 13 - 17%
  • TLC: One Spot
  • Sulphated Ash (dry basis): 0.1%

Applications:

Yeast Cell Culture; Gene Expression

TRUSTED_SUSTAINABILITY
Specifications

Specifications

15° to 30°C
100 g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
MP Biomedicals™ D-(+)- RAFFINOSE PENTAHYDRATE (POWDER) > 100g

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.