Learn More
MP Biomedicals™ CIRCLEGROW™, Powder
Shop All MP Biomedicals ProductsDescription
Perfect for overnight or weekend cultures, CIRCLEGROW Medium is a super-rich bacterial growth medium that produces up to three times more plasmid DNA than traditional growth medium. CIRCLEGROW Medium consists of a protein and amino acid source, biological buffer, and a complex carbon source that gets metabolized at a much slower rate than glucose, thus prolonging the cell cycle and preventing cell death during the log phase of growth.
- Cells grown with CIRCLEGROW Medium remain healthy for more than two days and continue to divide slowly after the log phase of the growth has been completed
- Reduce hands-on work and preparation time
- Cultures do not need to be constantly monitored by O.D. readings due to the incremental post-log cell growth
- Obtain high yields of plasmid DNA from smaller culture volumes, and confidently leave cultures overnight or over the weekend
- Capsules are the most convenient form of this unique growth media
- Pre-mixed CIRCLEGROW is also available in single-use pouches, capsules and sterile liquid
- MP113000165: Includes ten 20 g pouches. Add contents of one pouch to 500mL of purified water.
- MP113000175: Includes ten 40g pouches. Add contents of one pouch to 1L of purified water.
- MP113000185: Includes one hundred 20g pouches. Add contents of one pouch to 500mL of purified water.
Specifications
Specifications
| Content And Storage | 15° to 30°C |
| Product Type | Bacterial Growth Medium |
| Sterility | Sterile |
| pH | 6.9 |
| Quantity | 227 g |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.