Learn More
R&D Systems™ Mouse Th1 Cell Multi-Color Flow Cytometry Kit
For flow cytometric analysis of human Th1 cells using four fluorochrome-conjugated antibodies
Brand:  R&D Systems™ FMC010
Additional Details : Weight : 0.09700kg
Product Code. 18751874
  • 947.06€ / 1 set
Estimated Shipment: 18-07-2024
to see stock.
Description
Description
Includes conjugated antibodies (and corresponding isotype controls): IFN-gamma-CFS (Clone 37895; rat IgG2A), IL-12 Rbeta2-APC (Clone 305719; mouse IgG1), T-bet-PerCP (Clone 525803; mouse IgG1), CD4-PE (Clone GK1.5; rat IgG2B)
Specifications
Specifications
Store the unopened product at 2°C to 8°C. Do not use past expiration date.