missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mouse RARB (aa 429-448) Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP005
This item is not returnable.
View return policy
Description
Mouse Retinoic Acid (residues 429-448) Synthetic Peptide, PEP-005, from Invitrogen.
PEP-005 is a synthetic peptide whose sequence is from the C-terminal end of the mouse retinoic acid receptor beta (RAR*). The 21 amino acid sequence extends from amino acids 429 to 448 of RAR*. The sequence of PEP-005 is (amino to carboxy terminus): P S V S P S S V E N S G V S Q S P L L Q (The N-terminal cysteine has been added to facilitate conjugation and is not a part of the native mouse RAR* sequence)This peptide may be used for neutralization and control experiments with the monoclonal and polyclonal antibodies that reacts with this product and the human and mouse RAR*, catalog # MA1-811 and PA1-811. Using a solution with equal weights per unit volume of peptide and corresponding antibody will yield a solution with a large molar excess of peptide that is able to competitively bind the antibody.Reconstitute with 0.1 ml of distilled water.Specifications
218772 | |
purified | |
Mouse RARB (aa 429-448) | |
-20°C. Avoid freeze-thaw cycles. | |
Lyophilized | |
Control | |
Yes | |
Peptide | |
RUO - research use only |
Mouse | |
P22605 | |
0.5 mg/mL | |
Synthetic Peptide | |
peptide with no preservative | |
Rarb | |
No preservative | |
50 μg | |
P S V S P S S V E N S G V S Q S P L L Q |