missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-28B/IFN-lambda 3 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF1789
This item is not returnable.
View return policy
Description
IL-28B/IFN-lambda 3 Polyclonal specifically detects IL-28B/IFN-lambda 3 in Mouse samples. It is validated for Western Blot, Neutralization.
Specifications
| IL-28B/IFN-lambda 3 | |
| Polyclonal | |
| Unconjugated | |
| Q8CGK6 | |
| Ifnl3 | |
| E. coli-derived recombinant mouse IL-28B/IFN-lambda 3 Asp20-Val193 Accession # Q8CGK6 | |
| 100 μg | |
| Primary | |
| Detects IL-28B/IFN-lambda 3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 5% cross-reactivity with recombinant human IL-28A is observed. In Western blots, approximately 35% cross-reactivity (reducing conditions) and approximately 10% cross-reactivity (non-reducing conditions) with recombinant mouse IL-28A is observed. Neutralizes 100% of recombinant mouse IL-28A. | |
| Mouse | |
| IgG |
| Western Blot, Neutralization | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Neutralization 2-10 ug/mL | |
| interleukin-28B, IFN-lambda 3, IL28B, IL-28B, IL28C, interferon, lambda 3 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 282617 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction