Learn More
Invitrogen™ IFN alpha Mouse ELISA Kit
Sandwich ELISA Kit
Brand: Invitrogen™ 421201
Description
The Mouse Interferon alpha (Ms IFNα) ELISA quantitates Ms IFNα in mouse cell culture medium. The assay will exclusively recognize both natural and recombinant Ms IFNα. Principle of the method The Mouse IFNα solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples,standards,or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody,a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity,specificity,precision,and lot-to-lot consistency. See manual for more information on validation.
IFN alpha (Interferon alpha) is a Type I interferon belonging to a family of structurally related,species-specific proteins exclusive to vertebrates. The interaction of IFN-alpha with its receptor component result in the activation of a number of signaling pathways which are mainly involved in innate immune response against viral infection. In humans,the gene for IFN alpha is located on on chromosome 9. The type I interferons consist of 14 different alpha isoforms (subtypes with slightly different specificities),and single beta,omega,epsilon and kappa isoforms. Homologous molecules are found in many species,including rats and mice (and most mammals),and have been identified in birds,reptiles and fish species. IFN alpha is produced primarily by plasmacytoid dendritic cells,is a potent component of the anti-viral innate immune response and modulates adaptive immunity. Diseases associated with the dysfunction of IFN alpha include Hepatitis C and Hepatitis.Specifications
12.5-400pg/mL | |
HRP | |
ELISA Kit | |
No significant cross-reaction with mouse IFN-ß, IFN-g; human or rat IFN-a | |
Mouse | |
Ifna1 | |
<8% | |
HRP | |
Intra-assay CV ≤10%; inter-assay CV ≤ 10% | |
RUO | |
2°C to 8°C | |
1 hr. 20 min. |
Mouse IFN alpha ELISA Kit (Supernatant) | |
Colorimetric ELISA (TMB substrate) with Plates | |
Supernatant | |
96 microplate wells | |
Colorimetric Microplate Reader | |
<8% | |
Precoated 96-well Plate, 1 ea, Plate Sealers, 4 ea, Sample Buffer, 50mL, Stop Solution, 15mL, Wash Solution Concentrate, 50mL, TMB Substrate Solution, 15mL, Mouse IFN Alpha Standard 10,000pg/mL, 1 vial, Antibody Concentrate, 1 vial, Assay Diluent, 15mL, Concentrate Diluent, 15mL, HRP Conjugate Concentrate, 1 vial | |
Absorbance at 450nm | |
96 Tests | |
Supernatant, 100 μL | |
Mouse | |
27 hr. 15 min. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.