Learn More
R&D Systems™ Mouse GPR63 (NP_109658) VersaClone cDNA
R&D Systems™ Mouse GPR63 (NP_109658) VersaClone cDNA is a G protein-coupled receptor that may play a role in brain function.
Brand: R&D Systems™ RDC2203
This item is not returnable.
View return policy
Description
The Mouse GPR63 (NP_109658) VersaClone cDNA clone contains the complete ORF for GPR63, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The GPR63 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.Specifications
mGpr63 | |
10 μg at 0.2 μg/μL | |
1291bp | |
81491 | |
Mouse |
NP_109658 | |
cDNA is provided in 10 mM Tris-Cl | |
8.5 | |
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. | |
10 μg |