missing translation for 'onlineSavingsMsg'
Learn More

Proteintech™ Mouse Erythropoietin ELISA Kit

KE10031 is a solid phase sandwich Enzyme Linked-Immuno-Sorbent Assay (Sandwich ELISA). The Erythropoietin ELISA kit is to be used to detect and quantify protein levels of endogenous Erythropoietin.

Brand:  Proteintech™ KE10031-96T


Product Code. 16821626

  • 269.00€ / 96 tests
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • The assay recognizes mouse Erythropoietin. An antibody specific for Erythropoietin has been pre-coated onto the microwells.
  • The Erythropoietin protein in samples is captured by the coated antibody after incubation. Following extensive washing, another antibody of biotinylated specific for mouse Erythropoietin is added to detect the captured mouse Erythropoietin protein.
  • For signal development, Streptavidin-HRP is added, followed by Tetramethyl-benzidine (TMB) reagent.
  • Solution containing sulfuric acid is used to stop color development and the color intensity which is proportional to the quantity of bound protein is measurable at 450 nm with the correction wavelength set at 630 nm.
  • Erythropoietin (EPO) is a glycoprotein hormone that regulates the production of red blood cells and biosynthesis of hemoglobin.
  • The predominant expression of this gene shifts from the liver during fetal development to kidney in adults, and and the secreted protein will travel through the blood stream to reach to the bone marrow to stimulate hematopoietic stem cell differentiation to RBC.
  • EPO binds to the cognate EPO receptor (EPOR) on erythroid progenitor cells, thus preventing apoptosis and stimulating their differentiation and maturation into erythrocyte.
  • However, EPO protein and its receptors have also been shown to be cytoprotective in extra-hematopoietic tissues including the retina tissue.
  • A complete lack of erythropoietin causes embryonic lethal anemia in mice. The conditional inactivation of erythropoietin in adult mice results in a chronic, normocytic and normochromic anemia. 
Specifications

Specifications

BC119265
12 pg/mL
Sandwich ELISA, Double Antibody
Plasma, Serum
Mouse
Mouse
<10%
Blue ice
4.5 hr.
78.1 to 5000 pg/mL
ISO 13485
Coated ELISA Kit
DAB
13856
Solid
<10%
-20°C to +4°C
P07321
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 96TEST Mouse IL-4 ELISA Kit

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P234-Keep only in original container.
  • P264-Wash skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P330-Rinse mouth.
  • P362-Take off contaminated clothing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : Sulphuric Acid, Tetramethylbenzidine

SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.