missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Mouse EGF Recombinant Protein, PeproTech®
E. coli Recombinant Protein
80.00€ - 239.00€
Specifications
Accession Number | P01132 |
---|---|
For Use With (Application) | ELISA, Functional Assay, Western Blot |
Formulation | protein with no preservative |
Gene ID (Entrez) | 13645 |
Molecular Weight (g/mol) | 6 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17862843
|
Gibco™
315-09-100UG |
100 μg |
80.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17872513
|
Gibco™
315-09-1MG |
1 mg |
239.00€
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17852743
|
Gibco™
315-09-500UG |
500 μg |
194.00€
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
EGF (Epidermal growth factor) exerts its actions by binding to the EGF Receptor, a 170 kDa protein kinase. Activation of EGFR initiates diverse cellular pathways in response to toxic environmental stimuli, or to EGF binding to the receptor, the EGFR forms homo- or heterodimers with other family members. Each dimeric receptor complex initiates a distinct signaling pathway by recruiting different Src homology 2 (SH2) containing effector proteins. EGF is far and wide expressed in kidney, cerebrum, prostrate and salivary glands. EGF acts as a potent mitogenic factor and the phosphorylated receptor recruits adapter proteins like GRB2 that activates complex downstream signaling cascades. EGF activates at least 4 major downstream signaling cascades including the RAS-RAF-MEK-ERK, PI3 kinase-AKT, PLCgamma-PKC and STAT modules. Research studies suggest the protein may also play important role in activating the NF-kappa-B signaling cascade. Defects in the EGGF gene are the cause of hypomagnesemia type 4 and dysregulation has been associated with the growth and progression of certain cancers.Specifications
P01132 | |
protein with no preservative | |
6 | |
E. coli | |
RUO | |
AI790464; beta-urogastrone; EGF; Epidermal growth factor; epidermal growth factor (beta-urogastrone); epidermal growth factor precursor; H-EGF; HOMG4; Pro-epidermal growth factor; Pro-epidermal growth factor precursor (EGF); sb:eu639; URG; Urogastrone | |
Mouse | |
Recombinant | |
Lyophilized |
ELISA, Functional Assay, Western Blot | |
13645 | |
Mouse EGF | |
-20°C | |
<1 EU/μg | |
EGF | |
Unconjugated | |
NSYPGCPSSY DGYCLNGGVC MHIESLDSYT CNCVIGYSGD RCQTRDLRWW ELR | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |