missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mouse BMAL1 (aa 582-594) Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP075
This item is not returnable.
View return policy
Description
Mouse BMAL-1 (residues 582-594) Synthetic Peptide, PEP-075, from Invitrogen.
PEP-075 is a 13 amino acid synthetic peptide whose sequence corresponds to residues 582-594 of mouse BMAL-1 with an amino terminal cysteine added for carrier protein conjugation. The sequence of this peptide is (amino to carboxy terminus): (C)-D582-M-I-D-N-D-Q-G-S-S-S-P-S594This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and mouse BMAL-1, catalog # PA1-523. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∼70-fold) for competitive inhibition of antibody-protein binding reactions.Reconstitute with 0.1 ml of distilled water.Specifications
11865 | |
purified | |
Mouse BMAL1 (aa 582-594) | |
-20°C. Avoid freeze-thaw cycles. | |
Lyophilized | |
Control | |
Yes | |
Peptide | |
RUO - research use only |
Mouse | |
Q9WTL8 | |
0.5 mg/mL | |
Synthetic Peptide | |
peptide with no preservative | |
Arntl | |
No preservative | |
50 μg | |
(C) - D582 - M - I - D - N - D - Q - G - S - S - S - P - S594 |