Learn More
EPCAM, Mouse, Clone: ESA 214, Abnova™
Mouse monoclonal antibody raised against EPCAM.
Brand: Abnova MAB6195.500uL
Description
This gene encodes a carcinoma-associated antigen and is a member of a family that includes at least two type I membrane proteins. This antigen is expressed on most normal epithelial cells and gastrointestinal carcinomas and functions as a homotypic calcium-independent cell adhesion molecule. The antigen is being used as a target for immunotherapy treatment of human carcinomas. Mutations in this gene result in congenital tufting enteropathy. [provided by RefSeq
Specifications
| EPCAM | |
| Monoclonal | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:100) The optimal working dilution should be determined by the end user. | |
| EPCAM | |
| EPCAM | |
| Native purified EPCAM from small cell lung carcinoma. | |
| RUO | |
| 4072 | |
| Store at 4°C. | |
| IgG1 κ |
| Immunocytochemistry, Immunohistochemistry | |
| ESA 214 | |
| Mouse monoclonal antibody raised against EPCAM. | |
| In PBS, pH 7.4 (1% BSA, 0.05% sodium azide) | |
| 17-1A/323/A3/CD326/CO-17A/CO17-1A/EGP/EGP-2/EGP34/EGP40/ESA/Ep-CAM/GA733-2/HEA125/KS1/4/KSA/M4S1/MH99/MIC18/MK-1/MOC31/TACST-1/TACSTD1/TROP1/hEGP-2 | |
| Mouse | |
| 500 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.