Learn More
Monel™ 400 gauze, 40 mesh woven from 0.25mm (0.01 in.) dia. wire, Thermo Scientific™
33.47€ - 101.51€
Specifications
Chemical Name or Material | Monel™ 400 gauze |
---|---|
Name Note | 40 mesh woven from 0.25mm (0.01 in.) dia. wire |
Mesh Size | 40 Mesh |
Form | Gauze |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15406795
|
Thermo Scientific Alfa Aesar
045163.GT |
150 x 150 mm |
33.47€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15416795
|
Thermo Scientific Alfa Aesar
045163.HB |
300 x 300 mm |
101.51€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Monel™ 400 gauze | |
40 mesh woven from 0.25mm (0.01 in.) dia. wire | |
40 Mesh | |
Gauze |
Safety and Handling
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures