missing translation for 'onlineSavingsMsg'
Learn More

Molybdenum ICP Standard, 10000 mg/L Mo in NH3 4%, Honeywell Fluka™

Brand:  Honeywell Fluka E1232-100ML

Product Code. 16684024

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Honeywell Fluka™ analytical standards offer a growing range of high quality inorganic and organic reference materials for the European market produced according to ISO 9001. The products are suitable for a wide range of industries and applications and come complete with a detailed Certificate of Analysis (CoA) in every delivery, and are eligible for free expert technical support ensuring you have the information you need to quickly start your analysis and pass an audit.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Molybdenum ICP Standard
100 mL
Standards for ICP and ICP-MS
>11.5
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers