missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molybdenum AAS Standard, 1000 mg/L Mo in water, Honeywell Fluka™
34.05€ - 60.15€
Specifications
Chemical Name or Material | Molybdenum AAS Standard |
---|---|
Density | 0.999 g/cm3 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16664344
|
Honeywell Fluka
E4047-100ML |
100 mL | N/A | ||||||
16674344
|
Honeywell Fluka
E4047-500ML |
500 mL |
60.15€
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Honeywell Fluka™ analytical standards offer a growing range of high quality inorganic and organic reference materials for the European market produced according to ISO 9001. The products are suitable for a wide range of industries and applications and come complete with a detailed Certificate of Analysis (CoA) in every delivery, and are eligible for free expert technical support ensuring you have the information you need to quickly start your analysis and pass an audit.
Specifications
Molybdenum AAS Standard | |
0.999 g/cm3 |
Safety and Handling
missing translation for 'shelfLife' : 760