Learn More
Molecular Sieve Dehydrate with Indicator, Honeywell Fluka™
Rod Size 1/16 In., with Indicator for Drying Solvents
120.74€ - 399.05€
Specifications
Packaging | Poly bottle |
---|---|
Chemical Name or Material | Molecular Sieve Dehydrate Fluka with indicator for drying solvents |
UN Number | UN3077 |
Descrizione
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifica
Poly bottle | |
Molecular Sieve Dehydrate Fluka with indicator for drying solvents | |
UN3077 |
Sicurezza e movimentazione
P201-P261-P280-P284-P304 + P340-P308 + P313
missing translation for 'dotInformation' : 9/PGIII