Learn More
Invitrogen™ LIVE/DEAD™ Reduced Biohazard Cell Viability Kit #1, green & red fluorescence
Reduces risk associated with handling potential biohazards
Brand: Invitrogen™ L7013
334.68 EUR valid until 2024-09-28
Use promo code "21626" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21626” to get your promotional price
Description
The LIVE/DEAD Reduced Biohazard Cell Viability Kit No. 1 is a convenient and easy-to-use cell viability kit that is designed to reduce the risk associated with handling potential biohazards such as viral, bacterial or protozoan pathogens. The assay monitors viability as a function of the membrane integrity of the cell. Cells with a compromised membrane that are considered to be dead or dying will stain red, whereas cells with an intact membrane will stain green.
Cell Analysis, Cell Counting and Viability, Cell Counting, Viability, and Cryopreservation, Cell Culture, Cell Viability and Cytotoxicity, Cell Viability, Proliferation and Function, Flow Cytometry, Flow Cytometry Viability and Cytotoxicity Assays, Flow Cytometry of Cellular Processes
Order Info
Shipping Condition: Room temperature
Specifications
Eukaryotic Cells | |
Store in freezer at -5°C to -30°C and protect from light. | |
Fluorescence | |
Fluorescence Microscope, Flow Cytometer, Microplate Reader | |
Other Label(s) or Dye(s) | |
490 nm | |
LIVE/DEAD™ | |
Room Temperature |
Green, Red | |
LIVE/DEAD™ Reduced Biohazard Cell Viability Kit #1, green and red fluorescence | |
Viability Assay | |
Cell Viability Kit | |
516, 617 nm | |
Tube(s), 96-well plate, Slide(s) | |
1 kit |
For Research Use Only. Not for use in diagnostic procedures.