Learn More
Invitrogen™ BODIPY™ 558/568 C12 (4,4-Difluoro-5-(2-Thienyl)-4-Bora-3a,4a-Diaza-s-Indacene-3-Dodecanoic Acid)
The orange-red fluorescent fatty acid, BODIPY™ 558/568 C12 can be used as a synthetic precursor to a wide variety of fluorescent phospholipids.
Brand:  Invitrogen™ D3835
170.00 EUR valid until 2025-03-21
Use promo code "24111" to get your promotional price.
Product Code. 10493822
  • 255.00€ / 1mg
Explore more special offers
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Description
Description
The orange-red fluorescent fatty acid, BODIPY™ 558/568 C12 can be used as a synthetic precursor to a wide variety of fluorescent phospholipids. Unlike fluorophores like pyrene, DPH and NBD, BODIPY™ dyes are relatively environment insensitive and is fluorescent in both aqueous and lipid environments.
We recommend dissolving in high-quality anhydrous DMSO to attain a stock concentration up to 1 mM.
Order Info
Shipping Condition: Room Temperature
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Fatty Acids
Solid
559/568 nm
1 mg
BODIPY
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2896569 Certificate TypeCertificate of Analysis Date25/05/2024 Catalog NumberD3835
Lot Number2770864 Certificate TypeCertificate of Analysis Date27/12/2023 Catalog NumberD3835
Lot Number2581655 Certificate TypeCertificate of Analysis Date03/03/2023 Catalog NumberD3835
Lot Number2549237 Certificate TypeCertificate of Analysis Date11/11/2022 Catalog NumberD3835
Lot Number2507869 Certificate TypeCertificate of Analysis Date02/07/2022 Catalog NumberD3835
Product Title
Invitrogen™ BODIPY™ 558/568 C12 (4,4-Difluoro-5-(2-Thienyl)-4-Bora-3a,4a-Diaza-s-Indacene-3-Dodecanoic Acid) > 1 mg