Learn More
Epredia™ Modified Grocott' s Methenamine Silver (Periodic Acid) Stain Kit
Product Code. 12573647 Shop All Epredia Products
Click to view available options
Type:
Fast Green Stain Solution
Gold Chloride Solution (0.1%)
Methenamine/Borax
Periodic Acid Schiff
Silver Nitrate Solution (5%)
Silver Stain Kit, Periodic Acid
Sodium Thiosulfate Solution
Unit Size:
125mL
30mL
Each
Pack of 12
Product Code. 12573647
Brand: Epredia™ 88025
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Distinguish fungi, basement membranes and opportunistic organisms such as Pneumocystis Carinii in tissue sections with Epredia™ Modified Grocott's Methenamine Silver (Periodic Acid)
  • Fungi, basement membranes and opportunistic organisms, along with reticular fibers, are stained black
  • Background is stained green
TRUSTED_SUSTAINABILITY
Specifications
Type Sodium Thiosulfate Solution
Quantity 125mL