Learn More
Knick Elektronische™ Mobile pH Meter
Brand:  Knick Elektronische™ 902 pH
Product Code. 15315681
  • 1955.00€ / Each
Estimated Shipment: 13-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
The Portavo 902 pH Meter comes with: 
  • 4 AA batteries and pre-mounted quiver
  • Carrying strap
  • Quickstart instructions in various languages
  • Specific test report
  • Safety instructions
  • Data carrier with detailed user manuals
Specifications
Specifications
pH Meter, 4 AA batteries, Pre-mounted quiver, Carrying strap, Quickstart instructions, Specific test report, Data carrier with detailed user manuals
1300 to 1300
Yes
IP-66/67 with pressure compensation
500 g
Yes
Portavo 902 pH
Mobile pH Meter
-2 to 16
20°C to 120°C
DIN 19262
132 x 156 x 30 mm
3 Years
PA12 GF30 + TPE
0 to 95%, short-term condensing allowed
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products