missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ ML 315 hydrochloride

Low molecular weight, organic compound that may help regulate biological process.

229.00€ - 875.00€

Specifications

Product Type Inhibitor
Target Proteins Cdc2-like Kinase
CAS 1440251-53-5
Molecular Formula C18H13Cl2N3O2.HCl
Formula Weight 410.68g/mol
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18774539
View Documents
Tocris Bioscience™
5700/10
10 mg
229.00€
10mg
Please to purchase this item. Need a web account? Register with us today!
18714729
View Documents Promotion Details
Tocris Bioscience™
5700/50
50 mg
875.00€
50mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Inhibitor of cdc2-like kinases (Clk) and DYRK kinases (IC50 values are 68, 68 and 231 nM for Clk1, Clk4 and Clk2, respectively, and 282 and 1156 nM for Dyrk1A and Dyrk1B). Shows 20-fold selectivity for Clk4 over Dyrk1A. Stable in both mouse and human plasma. Moderately cell permeable.
Specifications

Specifications

Inhibitor
1440251-53-5
410.68g/mol
Cdc2-like Kinase Inhibitors
Cdc2-like Kinase
C18H13Cl2N3O2.HCl
Store at -20°C
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only