missing translation for 'onlineSavingsMsg'
Learn More

Mixed Calibration Standard 1 For Environmental Analysis, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Brand:  SPEX Certiprep MIXSTD1-500

 View more versions of this product

Product Code. 11971636

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »

Explore more special offers

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
This item is not returnable. View return policy

Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Pb500μg/mL
Se200μg/mL
Cd150μg/mL
Zn150μg/mL
Mn100μg/mL
Be50μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Mixed Calibration Standard 1
500 mL
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Components in 2% HNO3:Be    •50μg/mL
Cd    •150μg/mL
Pb    •500μg/mL
Mn    •100μg/mL
Se    •200μg/mL
Zn    •150μg/mL
Triple-leached LDPE bottle
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Environmental
2% HNO3
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product intended for laboratory use only