Learn More
Terracycle™ Mixed Beard Nets, Earplugs and Hair Nets Zero Waste Box™
GREENER_CHOICE
Recycle Mixed Beard Nets, Earplugs and Hair Nets using this Zero Waste Box system. Zero Waste Box includes signage, a built-in liner, shipping to and from your location, and processing of the materials.
Brand:  Terracycle™ CS46-M-BE
 View more versions of this product
Product Code. 17845925
  • 185.49€ / Each
Estimated Shipment: 09-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Use this Zero Waste Box to recycle:
  • Beard nets and hairnets
  • Disposable shoe covers
  • Earplugs
Do not send in:
  • Medical waste
    • This includes material contaminated with blood or bodily fluids and waste that originate from health care facilities, hospitals, home care, physicians' offices, dental offices, blood banks, veterinary clinics, research laboratories, etc.
  • Disposable masks or gloves
  • Lab coats or other garments
  • Hazardous waste (sharp, flammable, reactive, corrosive, ignitable, toxic, infectious or pathogenic) which presents a danger to the environment, or to people
  • Batteries, pressurized canisters and broken glass
Specifications
Specifications
15 kg
Beige, Green, Red, White
Single-use
Medium
Not Autoclavable
Recycling Box
33 lb.
30.5 x 30.5 x 84 cm
Yes
Non-sterile
Cardboard, Plastic