missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris™ Mitochondrial fusion promoter M1
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
Mitochondrial fusion promoter M1 is a mitochondrial fusion promoter. Protects cells from mitochondrial fragmentation-associated cell death. Induces mitochondrial fusion in effector T cells imposing a memory T cell morphology. Improves cellular respiration and potentiates glucose-stimulated insulin-secretion in cholesterol-exposed pancreatic β cells. Mitochondrial fusion promoter M1 also enhances mitochondrial dynamics and transport velocity in axons of the sciatic nerve in vitro and enhances axon regeneration in the optic nerve in an in vivo model of nerve damage.
Specifications
Specifications
| Chemical Name or Material | 4-Chloro-2-(1-(2-(2,4,6-trichlorophenyl)hydrazono)ethyl)phenol |
| CAS | 219315-22-7 |
| Quantity | 25 mg |
| Molecular Formula | C14H10Cl4N2O |
| Formula Weight | 364.05 |
| Product Type | Mitochondrial Fusion Promoter |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction