missing translation for 'onlineSavingsMsg'
Learn More

IKA™ Ministar 20 Digital
GREENER_CHOICE

Easy-to-use stirrers that combine basic functionalities with a compact design.

Brand:  IKA™ 0025004885

835.14 EUR valid until 2024-12-31
Use promo code "21658" to get your promotional price.


Product Code. 16238651

  • 1149.00€ / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Compact design - The reduced design and the focus on the most essential aspects make the stirrers reliable laboratory companions
  • Ease of operation - Operation takes place using a stable rotary knob
  • The display clearly shows the rotational speed
  • Rapid updates - Regular software updates can be undertaken rapidly and simply via the USB interface
Specifications

Specifications

50 to 2000 rpm
LED
60 w
Brushless DC
20 Ncm
1.56 kg
15 L
100/240 V
50/60 Hz
5°C to 40°C
10,000 mPas
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.