Learn More
HUBER Mineral Oil
High quality mineral oil based heat transfer fluid
293.00€ - 882.00€
Specifications
Color | Light Yellow |
---|---|
Temperature Range | 20°C to 190°C |
Type | Mineral Oil |
Temperature (Metric) Maximum | 190°C |
Viscosity | 40 mm2/sec. |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
11527374
|
HUBER
6156 |
20 L |
882.00€
20 L |
Estimated Shipment: 17-01-2025
Log in to see stock. |
|||||
10269651
|
HUBER
6155 |
5 L |
293.00€
5 L |
Estimated Shipment: 16-01-2025
Log in to see stock. |
|||||
Description
- Thermofluid P20.190.40
- Mineral oil
- Min. temp: 20°C
- Max. temp.: 190°C
- Viscosity at 25°C: 40mm²/s
Specifications
Light Yellow | |
Mineral Oil | |
40 mm2/sec. |
20°C to 190°C | |
190°C |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- minoil P20.190.40
Signal Word
- Danger
Hazard Category
- Aspiration toxicity Category 1
Hazard Statement
- H304-May be fatal if swallowed and enters airways.
Precautionary Statement
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : distallates (petroleum) solvent dewaxedheavy paraffinic