missing translation for 'onlineSavingsMsg'
Learn More

Milrinone, Tocris Bioscience™

Product Code. 18742484
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

PDE3 inhibitor

Potent cAMP phosphodiesterase inhibitor (IC50 = 56 nM for inhibition of PDE III). Has inotropic and vasodilator effects following oral or intravenous administration in vivo.

Chemical Identifiers

CAS 78415-72-2
Molecular Formula C12H9N3O
Molecular Weight (g/mol) 211.224
InChI Key PZRHRDRVRGEVNW-UHFFFAOYSA-N
Synonym milrinone, primacor, corotrope, corotrop, milrinona, milrinonum, milrinonum latin, milrinona spanish, milrila, unii-ju9yax04c7
PubChem CID 4197
ChEBI CHEBI:50693
IUPAC Name 6-methyl-2-oxo-5-pyridin-4-yl-1H-pyridine-3-carbonitrile
SMILES CC1=C(C=C(C(=O)N1)C#N)C2=CC=NC=C2

Specifications

Quantity 50 mg
Formula Weight 211.22
Percent Purity >98%
Chemical Name or Material Milrinone
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'