missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Midkine Antibody (2284A), PerCP, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB2581C
This item is not returnable.
View return policy
Description
Midkine Monoclonal antibody specifically detects Midkine in Human samples. It is validated for Flow Cytometry (Intracellular Staining), CyTOF
Specifications
| Midkine | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry (Intracellular Staining), CyTOF | |
| 2284A | |
| Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| Amphiregulin-associated protein, Midgestation and kidney protein, midkine, midkine (neurite growth-promoting factor 2), MK1, MKARAP, NEGF2FLJ27379, Neurite outgrowth-promoting factor 2, Neurite outgrowth-promoting protein | |
| E. coli-derived recombinant human Midkine., Lys23-Asp143, Accession # P21741 | |
| 0.1 mL | |
| Biologically Active Proteins, Cell Cycle and Replication | |
| 4192 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction