Learn More
Epredia™ Microm™ Cryostats Heat Extractor
Product Code. 12023059 Shop All Epredia Products
Click to view available options
For Use With (Equipment):
HM 525
HM 550 and 560
Unit Size:
Each
Product Code. 12023059
Brand: Epredia™ 524490
Use Epredia™ Microm™ Cryostats Heat Extractors with Epredia™ Cryostats.
  • Multipositional heat extractor is incorporated on a vertical shaft for even freezing and easy removal from specimen
  • Can be operated from any position on the freeze bar
  • Freezing time is shortened if freezing stage and tissue are placed on an active Peltier
TRUSTED_SUSTAINABILITY
Specifications
Product Type Heat Extractor
For Use With (Equipment) HM 525