missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One MICROLON™ 96-well ELISA Single-Break Strip Plates
For use in colorimetric immunoassay. Greiner Bio-One MICROLON™ 96-Well ELISA Single-Break Strip Plates can be broken off separately and number of tests performed can be adjusted to number of samples.
1195.00€ - 1265.00€
Specifications
Breakapart | Single break |
---|---|
Format | Strip Plate |
Array | 8 x 12 |
Material | Polystyrene |
No. of Wells | 96 |
Product Code | Brand | Color | Binding Property | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Binding Property | Price | Quantity & Availability | ||||
10063301
|
Greiner Bio-One
705070 |
Clear | Medium Binding |
1265.00€
Pack of 100 |
Estimated Shipment: 04-03-2025
Log in to see stock. |
||||
10746732
|
Greiner Bio-One
705071 |
Clear | High Binding |
1195.00€
Pack of 100 |
Estimated Shipment: 04-03-2025
Log in to see stock. |
||||
Description
- Free of detectable DNase, RNase, human DNA Non-pyrogenic
- Individual wells can be pipetted without leaving residue
- Flat bottom enables precise optical measurements
- Suitable for transmission measurements
- Specifica
Specifications
Single break | |
8 x 12 | |
96 | |
C | |
Colorimetric immunoassay | |
127.76 x 85.48 x 14.5 mm (5.02 x 3.36 x 0.5 in.) |
Strip Plate | |
Polystyrene | |
Non-sterile | |
Yes | |
Yes |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title