Learn More
3M™ Microbial Luminescence System MLSII
A rapid microbial detection system using ATP bioluminescence in a microwell based System to detect the presence of microbial ATP in Ultra Heat Treated (UTH) and Extended Shelf Life (ESL) dairy and dairy end products.
Brand:  3M™ 7000032756
Additional Details : Weight : 0.48500kg
Product Code. 17218530
  • 44387.50€ / 1 set
Estimated Shipment: 02-09-2024
to see stock.
Add to basket
Description
Description
  • Microplate, 5 injector-dispenser luminometer designed for microbial detection.
  • Results available approximately 48 hours sooner than traditional methods.
  • Reduces stock holding requirements and improves responsiveness to customers.
  • Time saved improves cash flow.
  • Allows quality performance to be tracked over time.
Specifications
Specifications
1