Learn More
SI Analytics™ Micro-Ubbelohde Viscometers (DIN)
Micro-Ubbelohde viscometers are calibrated for manual measurements
Brand:  SI Analytics™ 285401017
Product Code. 10662642
  • 528.00€ / Each
Estimated Shipment: 23-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Calibrated for manual measurement
  • DIN
  • Type No. 536 13
  • Capillary No. Ic
  • Constant K = 0.03
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Ubbelohde Micro Viscometer
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products