Learn More
SI Analytics™ Micro-Ostwald Viscometers
Micro-Ostwald viscometers are calibrated for automatic measurement
Brand:  SI Analytics™ 285404322
Product Code. 10102081
  • 392.00€ / Each
Estimated Shipment: 31-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Calibrated for automatic measurement
  • Type No. 517 20
  • Capillary No. II
  • Constant K = 0.1
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Micro-Ostwald Viscometer
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers