Learn More
Mettler Toledo™ Peel-Off Self-Adhesive Labels for P50 Line Thermal Printers
For use with P50 Line Thermal Printers.
141.00€
Specifications
For Use With (Equipment) | P-56RUE and P-58RUE Thermal Printers |
---|---|
Type | Peel-off Self-adhesive Labels |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15403563
|
Mettler Toledo™
30094724 |
10 Pieces |
141.00€
10 rolls |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Peel-off self-adhesive labels
- Accessory for P-56RUE and P-58RUE Thermal printers
- White in color
- Upto 10 years durability
- 58mm
- Ten pieces
- Accessory for P-56RUE and P-58RUE Thermal printers
- White in color
- Upto 10 years durability
- 54 x 18mm
- Six pieces
Specifications
P-56RUE and P-58RUE Thermal Printers | |
Peel-off Self-adhesive Labels |