Learn More
Metro™ Starsys™ Overhead Storage Unit
Microban™ antimicrobial protection prevents growth of stains and odor-causing bacteria.
Brand: Metro™ SXRDOH27P2C
This item is not returnable.
View return policy
Description
- Built on same widths as Starsys base units utilizing very similar construction
- Insures that wall cabinets align with lower product configuration in application
- All overhead cabinets include corrugated polymer inner panels in their assembly as well as clean-design back panel, molded with cosmetic side facing inside of cabinet, creating much cleaner appearance - especially when used without doors or in conjunction with clear door
- Necessary wall mount rails/hardware are included with each wall cabinet
- Microban™ antimicrobial protection inhibits growth of stain and odor-causing bacteria on Starsys, keeping surface areas cleaner between cleanings
Specifications
ISO 9001-9002 | |
Fixed Window | |
Doors with View, Double Width, Solid Shelves | |
2 | |
41.3 in. | |
13.8 in. | |
Polymer | |
41.3 in. |
13.8 x 41.3 x 29.5 in. | |
29.5 in. | |
2 | |
Solid Shelf | |
Taupe | |
29.5 in. | |
Overhead Storage Unit |