Learn More
Metro™ Stainless Lab Worktable, Stainless Top/Backsplash and Solid HD Shelf
Product Code. 15977111
Click to view available options
Dimensions (L x W x H):
29.75 x 30 x 35 in.
35.75 x 30 x 35 in.
47.75 x 30 x 35 in.
59.75 x 30 x 35 in.
Includes:
Backsplash, Solid HD Shelf
Backsplash, Solid HD Shelf, Adapter Insert
Product Type:
Mobile Table
Stationary Table
Unit Size:
Each
Product Code. 15977111
Brand: Metro™ LTS60S
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Provide a safer workstation for your staff. Metro Stainless Lab Worktable features a backsplash, solid shelf, corrosion-proof performance and stainless steel construction throughout.
  • Easily cleaned smooth surfaces
  • Mobile units can be easily moved to clean behind table area
  • Wide range of accessories and caster options available
Order Info
Casters must be ordered separately.
Specifications
Product Type Stationary Table
Dimensions (L x W x H) 59.75 x 30 x 35 in.
For Use With (Application) Ideal for lab
Height (English) 35 in.
Height (Metric) 89 cm
Includes Backsplash, Solid HD Shelf
Length (English) 59.75 in.
Length (Metric) 151.7 cm
Load Bearing Capacity (English) 600 lb.
Load Bearing Capacity (Metric) 273 kg
Width (English) 30 in.
Width (Metric) 76.2 cm
Material Stainless Steel