missing translation for 'onlineSavingsMsg'
Learn More

Methyl thiazolidine-2-carboxylate hydrochloride, 98%, Thermo Scientific Chemicals

55.80€ - 190.00€

Chemical Identifiers

CAS 50703-06-5
Molecular Formula C5H10ClNO2S
Molecular Weight (g/mol) 183.65
MDL Number MFCD00068022
InChI Key YEFOSJYXVLNYSL-UHFFFAOYNA-N
Synonym 2-thiazolidinecarboxylic acid methyl ester, 2-thiazolidinecarboxylic acid, methyl ester, thiazolidine-2-carboxylic acid methyl ester, methyl thiazolidine-2-carboxylate, +-thiazolidine-2-carboxylic acid methyl ester, +/--thiazolidine-2-carboxylic acid methyl ester, 2-thiazolidinecarboxylic acid, methyl ester 6ci,8ci,9ci
PubChem CID 2737507
IUPAC Name methyl 1,3-thiazolidine-2-carboxylate
SMILES [H+].[Cl-].COC(=O)C1CSCN1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11382966
View Documents Promotion Details
Thermo Scientific Alfa Aesar
L10740.03
1 g
55.80€
1g
Please to purchase this item. Need a web account? Register with us today!
11392966
View Documents Promotion Details
Thermo Scientific Alfa Aesar
L10740.06
5 g
190.00€
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

50703-06-5
183.65
YEFOSJYXVLNYSL-UHFFFAOYNA-N
2737507
[H+].[Cl-].COC(=O)C1CSCN1
C5H10ClNO2S
MFCD00068022
2-thiazolidinecarboxylic acid methyl ester, 2-thiazolidinecarboxylic acid, methyl ester, thiazolidine-2-carboxylic acid methyl ester, methyl thiazolidine-2-carboxylate, +-thiazolidine-2-carboxylic acid methyl ester, +/--thiazolidine-2-carboxylic acid methyl ester, 2-thiazolidinecarboxylic acid, methyl ester 6ci,8ci,9ci
methyl 1,3-thiazolidine-2-carboxylate

Specifications

50703-06-5
C5H10ClNO2S
3628640
YEFOSJYXVLNYSL-UHFFFAOYNA-N
methyl 1,3-thiazolidine-2-carboxylate
2737507
98%
∽158°C (decomposition)
MFCD00068022
2-thiazolidinecarboxylic acid methyl ester, 2-thiazolidinecarboxylic acid, methyl ester, thiazolidine-2-carboxylic acid methyl ester, methyl thiazolidine-2-carboxylate, +-thiazolidine-2-carboxylic acid methyl ester, +/--thiazolidine-2-carboxylic acid methyl ester, 2-thiazolidinecarboxylic acid, methyl ester 6ci,8ci,9ci
[H+].[Cl-].COC(=O)C1CSCN1
183.65
183.66
Methyl thiazolidine-2-carboxylate hydrochloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 256-726-5

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only