Learn More
Methyl nitroacetate, 97+%, Thermo Scientific Chemicals
75.80€ - 309.00€
Chemical Identifiers
CAS 2483-57-0
Molecular Formula C3H5NO4
Molecular Weight (g/mol) 119.076
MDL Number MFCD00075480
InChI Key ALBSWLMUHHZLLR-UHFFFAOYSA-N
Synonym methyl nitroacetate, nitroacetic acid methyl ester, acetic acid, nitro-, methyl ester, methylnitroacetate, methyl nitrozcetate, methyl nitro acetate, zlchem 149, methyl a-nitroacetate, methyl 2-nitroethanoate, acmc-1clk8
PubChem CID 17206
IUPAC Name methyl 2-nitroacetate
SMILES COC(=O)C[N+](=O)[O-]
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11379555
View Documents Promotion Details
Thermo Scientific Alfa Aesar
L18950.04
2 g
75.80€
2g
11389555
View Documents Promotion Details
Thermo Scientific Alfa Aesar
L18950.09
10 g
309.00€
10g
Description
Description
Methyl nitroacetate is used in the synthesis of aromatic and heteroaromatic linear (E)-alfa-nitro-arylpentenoates. It is also used to prepare phenyliodonium ylide. Further, it is utilized in the highly enantioselective and diastereoselective copper(I)-catalyzed cyclopropanation of alkenes. In addition to this, it is employed in the production of methyl (Z)-2-nitro-3-(4-nitrophenyl)-2-propenoate through reaction with 4-nitrobenzylideneaniline.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methyl nitroacetate is used in the synthesis of aromatic and heteroaromatic linear (E)-alfa-nitro-arylpentenoates. It is also used to prepare phenyliodonium ylide. Further, it is utilized in the highly enantioselective and diastereoselective copper(I)-catalyzed cyclopropanation of alkenes. In addition to this, it is employed in the production of methyl (Z)-2-nitro-3-(4-nitrophenyl)-2-propenoate through reaction with 4-nitrobenzylideneaniline.
Solubility
Miscible with ethanol, ether and most organic solvents.
Notes
Light sensitive. Incompatible withstrong bases.
Specifications
Chemical Identifiers
2483-57-0
119.076
ALBSWLMUHHZLLR-UHFFFAOYSA-N
17206
COC(=O)C[N+](=O)[O-]
C3H5NO4
MFCD00075480
methyl nitroacetate, nitroacetic acid methyl ester, acetic acid, nitro-, methyl ester, methylnitroacetate, methyl nitrozcetate, methyl nitro acetate, zlchem 149, methyl a-nitroacetate, methyl 2-nitroethanoate, acmc-1clk8
methyl 2-nitroacetate
Specifications
2483-57-0
195°C to 198°C
Ester-like
O2NCH2CO2CH3
MFCD00075480
Light sensitive
Miscible with ethanol,ether and most organic solvents.
COC(=O)C[N+](=O)[O-]
119.076
119.08
Methyl nitroacetate
1.294
105°C (221°F)
C3H5NO4
1.426
1758670
methyl nitroacetate, nitroacetic acid methyl ester, acetic acid, nitro-, methyl ester, methylnitroacetate, methyl nitrozcetate, methyl nitro acetate, zlchem 149, methyl a-nitroacetate, methyl 2-nitroethanoate, acmc-1clk8
ALBSWLMUHHZLLR-UHFFFAOYSA-N
methyl 2-nitroacetate
17206
98%
Videos
Safety and Handling
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 219-622-0
missing translation for 'rtecsNumber' : AJ1093000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures