missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl acetate, CHROMASOLV™, for HPLC, ≥99.8%, Honeywell Riedel-de Haën™
Click to view available options
Quantity:
250 mL
2.5 L
Description
- For HPLC and organic synthesis
- High-purity, multipurpose solvents
- High lot-to-lot consistency
Specifications
Specifications
CAS Max % | ≤100.0000% |
Melting Point | -98°C (literature) |
Density | 0.934 g/mL at 25 °C |
Boiling Point | 57°C to 58°C |
Flash Point | −12.99°C |
Packaging | Glass Bottle |
Linear Formula | CH3COOCH3 |
Quantity | 2.5 L |
UN Number | UN1231 |
Beilstein | 1736662 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'