missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-hydroxyphenylacetate, 98+%, Thermo Scientific Chemicals

Product Code. 11470737
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14199-15-6
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00002387
InChI Key XGDZEDRBLVIUMX-UHFFFAOYSA-N
Synonym methyl 4-hydroxyphenylacetate, methyl 2-4-hydroxyphenyl acetate, 4-hydroxyphenylacetic acid methyl ester, benzeneacetic acid, 4-hydroxy-, methyl ester, methyl 4-hydroxyphenyl acetate, 4-hydroxyphenylacetic acid methl ester, acetic acid, p-hydroxyphenyl-, methyl ester, p-hydroxyphenylacetic acid methyl ester, 4-hydroxybenzeneacetic acid methyl ester, 4-hydroxy-phenyl-acetic acid methyl ester
PubChem CID 518900
ChEBI CHEBI:68078
IUPAC Name methyl 2-(4-hydroxyphenyl)acetate
SMILES COC(=O)CC1=CC=C(C=C1)O

Specifications

Melting Point 55°C to 58°C
Boiling Point 163°C (5 mmHg)
Quantity 25 g
Beilstein 2087538
Formula Weight 166.18
Percent Purity ≥98%
Chemical Name or Material Methyl 4-hydroxyphenylacetate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'