Learn More
Methyl 2-hydroxy-5,6,7,8-tetrahydronaphthalene-1-carboxylate, 97%, Thermo Scientific Chemicals
Product Code. 15437865
Click to view available options
Quantity:
250 mg
1 g
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 59604-96-5
Molecular Formula C12H14O3
Molecular Weight (g/mol) 206.24
MDL Number MFCD09258894
InChI Key FGZPUQMMDVJQQN-UHFFFAOYSA-N
Synonym 6-hydroxytetralin-5-carboxylic acid methyl ester, methyl 2-hydroxy-5,6,7,8-tetrahydro-1-naphthoate, 2-hydroxy-5,6,7,8-tetrahydro-naphthalene-1-carboxylic acid methyl ester, 2-hydroxy-5,6,7,8-tetrahydronaphthalene-1-carboxylic acid methyl ester
PubChem CID 11600901
IUPAC Name methyl 2-hydroxy-5,6,7,8-tetrahydronaphthalene-1-carboxylate
SMILES COC(=O)C1=C(O)C=CC2=C1CCCC2
Specifications
Melting Point 40°C to 43°C
Quantity 1 g
Formula Weight 206.24
Percent Purity 97%
Chemical Name or Material Methyl 2-hydroxy-5,6,7,8-tetrahydronaphthalene-1-carboxylate